Skip to Content
Merck
All Photos(1)

Key Documents

903965

Sigma-Aldrich

Trimethylboroxine

50% THF solution

Synonym(s):

2,4,6-Trimethyl-1,3,5,2,4,6-trioxatriborinane, 2,4,6-Trimethylboroxine, Methaneboronic anhydride, Trimethyl-1,3,5,2,4,6-trioxatriborinane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H9B3O3
CAS Number:
Molecular Weight:
125.53
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.22

form

liquid

refractive index

n/D 1.3880

density

0.89962 g/mL

SMILES string

B1(OB(OB(O1)C)C)C

InChI

1S/C3H9B3O3/c1-4-7-5(2)9-6(3)8-4/h1-3H3

InChI key

GBBSAMQTQCPOBF-UHFFFAOYSA-N

Application

Trimethylboroxine (TMB) is a cyclic anhydride of methyl-boronic acid. It can be used as a:
  • Methylating agent for the methylation of various aromatic halides and C(sp3)−H bonds using palladium catalyst.
  • Reagent in the preparation of polymer supported CBS (Corey, Bakshi, and Shibata) catalysts.

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-5.8 °F

Flash Point(C)

-21 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oxazaborolidines as functional monomers: ketone reduction using polymer-supported Corey, Bakshi, and Shibata catalysts.
Price MD, et al.
The Journal of Organic Chemistry, 67(23), 8086-8089 (2002)
Jacob B Geri et al.
Journal of the American Chemical Society, 139(29), 9811-9814 (2017-07-14)
We present a strategy to rationally prepare CF3- transfer reagents at ambient temperature from HCF3. We demonstrate that a highly reactive CF3- adduct can be synthesized from alkali metal hydride, HCF3, and borazine Lewis acids in quantitative yield at room
Profound Methyl Effects in Drug Discovery and a Call for New C−H Methylation Reactions.
Schoenherr H and Cernak T
Angewandte Chemie (International ed. in English), 52(47), 12256-12267 (2013)
Practical methylation of aryl halides by Suzuki-Miyaura coupling.
Gray M, et al.
Tetrahedron Letters, 41(32), 6237-6240 (2000)
Anne Rietz et al.
Journal of medicinal chemistry, 60(11), 4594-4610 (2017-05-10)
Spinal muscular atrophy (SMA) is the leading genetic cause of infant death. We previously developed a high-throughput assay that employs an SMN2-luciferase reporter allowing identification of compounds that act transcriptionally, enhance exon recognition, or stabilize the SMN protein. We describe

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service