901775
Tungsten disulfide suspension
Synonym(s):
2D-Tungsten disulfide, 2D-Tungsten disulfide dispersion with nonionic surfactant, 2D-Tungsten sulfide dispersion, Tungsten sulfide
About This Item
Recommended Products
description
thickness < 3 layers
50-300 nm
form
liquid
concentration
5 mg/mL in H2O
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
Storage Class Code
12 - Non Combustible Liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service