Skip to Content
Merck
All Photos(1)

Key Documents

901588

Sigma-Aldrich

1,3-Propanesultone solution

1 M in THF

Synonym(s):

1,2-Oxathiolane 2,2-dioxide, 3-Hydroxypropanesulfonic acid γ-sultone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C3H6O3S
CAS Number:
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.22

form

liquid

concentration

1 M in THF

refractive index

n/D 1.4144

density

0.9266 g/mL

InChI

1S/C3H6O3S/c4-7(5)3-1-2-6-7/h1-3H2

InChI key

FSSPGSAQUIYDCN-UHFFFAOYSA-N

Application

1,3-Propanesultone has been used in:
  • preparation of poly[2-ethynyl-N-(propylsulfonate)pyridinium betaine], a new ionic conjugated polymer
  • preparation of novel poly(4-vinylpyridine) supported acidic ionic liquid catalyst
  • preparation of poly((2-(dimethylamino)ethyl methacrylate)-co-3-dimethyl(methacryloyloxyethyl)ammonium propanesulfonate)-coated mesoporous silica nanoparticles
  • sulfonation of surface of self-assembled nanoporous silica colloidal crystalline films comprised of 184nm-diameter silica spheres
Valuable building block or solvent offered as a solution in tetrahydrofuran for more convenient handling.

related product

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B - Eye Dam. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-2.0 °F

Flash Point(C)

-18.88 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Junke Wang et al.
Ultrasonics sonochemistry, 21(1), 29-34 (2013-06-12)
A novel poly(4-vinylpyridine) supported acidic ionic liquid catalyst was synthesized by the reaction of 4-vinylpyridine with 1,3-propanesultone, followed by the polymerization and the addition of the heteropolyacid. Due to the combination of polymer features and ionic liquid, it acted as
Yeong-Soon Gal et al.
Journal of nanoscience and nanotechnology, 14(7), 5480-5484 (2014-04-25)
A new ionic conjugated polymer was prepared by the activated polymerization of 2-ethynylpyridine with the ring-opening of 1,3-propanesultone without any additional initiator or catalyst. This polymer was characterized by various instrumental methods to have conjugated polymer backbone system with pendant
Stephen D Dertinger et al.
Environmental and molecular mutagenesis, 52(9), 748-755 (2011-11-05)
An international collaborative trial was established to systematically investigate the merits and limitations of a rat in vivo Pig-a gene mutation assay. The product of this gene is essential for anchoring CD59 to the plasma membrane, and mutations in this
D K Torous et al.
Mutation research, 465(1-2), 91-99 (2000-03-10)
Micronuclei (MN) are routinely enumerated in mouse peripheral blood to index genotoxicity. Recent data from the Collaborative Study Group for the Micronucleus Test (CSGMT) [CSGMT (The Collaborative Study Group for the Micronucleus Test), Evaluation of the rat micronucleus test with
Hermann M Bolt et al.
Toxicology letters, 151(1), 251-254 (2004-06-05)
1,3-Propane sultone is directly alkylating, genotoxic and carcinogenic. In rats, it induces local and systemic tumours at multiple target sites. Preponderant systemic tumours occur at the central nervous system, especially gliomas. Other localisations include the mammary gland, the intestine, the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service