Skip to Content
Merck
All Photos(1)

Key Documents

901249

Sigma-Aldrich

Pyruvic-13C3 acid

endotoxin tested, ≥99 atom % 13C, ≥95% (CP)

Synonym(s):

α-Ketopropionic acid-13C3, 2-Oxopropionic acid-13C3

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
13CH313CO13COOH
CAS Number:
Molecular Weight:
91.04
MDL number:
UNSPSC Code:
12352115

isotopic purity

≥99 atom % 13C

Assay

≥95% (CP)

form

liquid

bp

165 °C (lit.)

mp

11-12 °C (lit.)

suitability

endotoxin tested

mass shift

M+3

SMILES string

[13CH3][13C](=O)[13C](O)=O

InChI

1S/C3H4O3/c1-2(4)3(5)6/h1H3,(H,5,6)/i1+1,2+1,3+1

InChI key

LCTONWCANYUPML-VMIGTVKRSA-N

Looking for similar products? Visit Product Comparison Guide

Application

For R&D use only. Not for drug, household, or other uses. Any other use is the sole responsibility of the end user.




Endotoxin testing is done on the bulk form of the product before subdivision packaging. This bulk test does not guarantee that the subdivision or repackaged aliquot is endotoxin-free when it is received or used by the customer and does not imply suitability for any particular purpose. If the product must be endotoxin-free for the intended application, the product should be tested prior to actual use.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1C

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

179.6 °F - closed cup

Flash Point(C)

82 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service