Skip to Content
Merck
All Photos(3)

Key Documents

900711

Sigma-Aldrich

Carbon nanotube, single-walled

≥98% carbon basis, avg. no. of layers, 1

Synonym(s):

ZEONANO, SWCNT, SWNT, Single wall carbon nanotube

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352103
NACRES:
NA.23

Quality Level

Assay

≥98% carbon basis
<1% (ash)

form

powder or flakes

feature

avg. no. of layers 1

L

300-500 μm , in forest

avg. diam.

3-5 nm

surface area

800-1000 m2/g , by BET

impurities

500 ppm Fe (max)

color

black

mp

3652-3697 °C (lit.)

density

1.7-1.9 g/cm3 at 25 °C (lit.)

SMILES string

[C]

Looking for similar products? Visit Product Comparison Guide

Application

  • Electrode in power storage devices.
  • Additives in structural materials.
  • Flexible electronic materials.

Preparation Note

Super-growth (SG) method.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lorena González-Legarreta et al.
Analytical methods : advancing methods and applications, 12(46), 5642-5647 (2020-11-14)
Understanding the biological effects triggered by nanomaterials is crucial, not only in nanomedicine but also in toxicology. The dose-response relation is relevant in biological tests due to its use for determining appropriate dosages for drugs and toxicity limits. Carbon nanotubes

Articles

Single-Walled Carbon Nanotubes synthesized by the Super-Growth Method & their properties & applications, including dispersing SGCNTs, SGCNT-polymer composites & SGCNT-metal composites are discussed.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service