88720
(S)-1-[(RP)-2-(Diphenylphosphino)ferrocenyl]ethyldi-tert-butylphosphine
≥97%
Synonym(s):
(2S)-1-[(1S)-1-[Bis(1,1-dimethylethyl)phosphino]ethyl]-2-(diphenylphosphino)ferrocene (acc to CAS), Josiphos SL-J002-2
About This Item
Recommended Products
Assay
≥97%
form
powder
optical activity
[α]20/D +410±15°, c = 0.5 in chloroform
optical purity
ee: ≥99%
functional group
phosphine
SMILES string
[Fe].[CH]1[CH][CH][CH][CH]1.C[C@@H]([C]2[CH][CH][CH][C]2P(c3ccccc3)c4ccccc4)P(C(C)(C)C)C(C)(C)C
InChI
1S/C27H35P2.C5H5.Fe/c1-21(29(26(2,3)4)27(5,6)7)24-19-14-20-25(24)28(22-15-10-8-11-16-22)23-17-12-9-13-18-23;1-2-4-5-3-1;/h8-21H,1-7H3;1-5H;/t21-;;/m0../s1
InChI key
STAGMXYQPGLLTD-FGJQBABTSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Packaging
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service