Skip to Content
Merck
All Photos(1)

Key Documents

88545

Sigma-Aldrich

1,1′-Thiocarbonyldiimidazole

≥95.0% (S)

Synonym(s):

TCDI

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H6N4S
CAS Number:
Molecular Weight:
178.21
Beilstein:
609349
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥95.0% (S)

form

powder

reaction suitability

reaction type: Carbonylations

mp

98-102 °C (lit.)

functional group

thiourea

storage temp.

2-8°C

SMILES string

S=C(n1ccnc1)n2ccnc2

InChI

1S/C7H6N4S/c12-7(10-3-1-8-5-10)11-4-2-9-6-11/h1-6H

InChI key

RAFNCPHFRHZCPS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • 1,1′-Thiocarbonyldiimidazole (TCDI) is generally used as a reagent to carry out the deoxygenation of vicinal diols by forming cyclic thionocarbonate in Corey-Winter alkene synthesis.
  • It is used as a reagent in the enantioselective total synthesis of (+)-hapalindole Q, 12-epi-fischerindole U and welwitindolinone A.
  • It is also used in the preparation of trithiocarbonates, xanthates, and dithiocarbamates as chain transfer agents for the reversible addition fragmentation chain transfer and macromolecular design (RAFT/ MADIX) polymerization.

Other Notes

Reagent for the synthesis of thioamides; Used in a modified Corey-Winter olefin synthesis; Preparation of thiocarbamates from alcohols which undergo radical reactions with Bu3SnH-AIBN

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Direct coupling of indoles with carbonyl compounds: short, enantioselective, gram-scale synthetic entry into the hapalindole and fischerindole alkaloid families.
Baran PS and Richter JM
Journal of the American Chemical Society, 126(24), 7450-7451 (2004)
1, 1?-Thiocarbonyldiimidazole.
e-EROS Encyclopedia of Reagents for Organic Synthesis (2001)
Enantiospecific total synthesis of the hapalindoles, fischerindoles, and welwitindolinones via a redox economic approach.
Richter JM, et al.
Journal of the American Chemical Society, 130(52), 17938-17954 (2008)
Selective one-pot synthesis of trithiocarbonates, xanthates, and dithiocarbamates for use in RAFT/MADIX living radical polymerizations.
Wood MR, et al.
Organic Letters, 8(4), 553-556 (2006)
S. Hanessian et al.
Canadian Journal of Chemistry, 65, 1859-1859 (1987)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service