Skip to Content
Merck
All Photos(1)

Key Documents

87249

Sigma-Aldrich

Tetradodecylammonium bromide

≥99.0% (AT)

Synonym(s):

Tetralaurylammonium bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)10CH2]4N(Br)
CAS Number:
Molecular Weight:
771.22
Beilstein:
5844282
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥99.0% (AT)

form

crystals

impurities

≤0.5% water

SMILES string

[Br-].CCCCCCCCCCCC[N+](CCCCCCCCCCCC)(CCCCCCCCCCCC)CCCCCCCCCCCC

InChI

1S/C48H100N.BrH/c1-5-9-13-17-21-25-29-33-37-41-45-49(46-42-38-34-30-26-22-18-14-10-6-2,47-43-39-35-31-27-23-19-15-11-7-3)48-44-40-36-32-28-24-20-16-12-8-4;/h5-48H2,1-4H3;1H/q+1;/p-1

InChI key

SMEFTBPJZGVAPK-UHFFFAOYSA-M

Related Categories

General description

Tetradodecylammonium bromide (TDAB) is a quaternary ammonium salt consisting of positively charged nitrogen atoms attached to a long alkyl chain. It is commonly used as a surfactant and phase-transfer catalyst.

Application

Tetradodecylammonium bromide can be used as a phase transfer catalyst in the synthesis of ethyl-lophine.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yusuke Tahara et al.
Sensors (Basel, Switzerland), 13(1), 1076-1084 (2013-01-18)
We have developed a new portable taste sensor with a lipid/polymer membrane and conducted experiments to evaluate the sensor's performance. The fabricated sensor consists of a taste sensor chip (40 mm × 26 mm × 2.2 mm) with working and
Tatsumi Mizuta et al.
The Analyst, 145(16), 5430-5437 (2020-07-15)
Herein, a fully lipophilic ionic liquid (IL) comprising a lipophilic fluorescein anion and a trihexyltetradecylphosphonium cation was synthesized and used as the plasticizer for a plasticized poly(vinyl chloride) (PVC) membrane optode. Systematic investigation of the alkyl chain length of the
Electrochemically controlling the size of gold nanoparticles
Huang CJ, et al.
Journal of the Electrochemical Society, 153(12), D193-D193 (2006)
Synthesis and structure of 1-ethyl-2, 4, 5-triphenyl-1H-imidazole (ethyl-lophine)
Zeitschrift fur Naturforschung B, 68(3), 245-249 (2013)
Magdalena Münster et al.
International journal of pharmaceutics, 529(1-2), 310-318 (2017-07-12)
The taste of pharmaceuticals strongly affects the compliance of patients. This study investigated the applicability of the electronic tongue and rodent brief-access taste aversion (BATA) model for the bitter compound praziquantel (PZQ) and taste masked liquid formulations for PZQ. In

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service