86341
TentaGel™ HL-PHB
extent of labeling: ~0.35 mmol/g loading
Synonym(s):
O-[4-(Hydroxymethyl)phenyl]polyethyleneglycol polymer-bound
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
form
powder
Quality Level
extent of labeling
~0.35 mmol/g loading
particle size
~75 μm
application(s)
peptide synthesis
storage temp.
2-8°C
Legal Information
TentaGel is a trademark of Rapp Polymere GmbH
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service