Skip to Content
Merck
All Photos(3)

Key Documents

851450

Sigma-Aldrich

D-(+)-Glucuronic acid γ-lactone

≥99%

Synonym(s):

D-(+)-Glucurono-6,3-lactone, D-Glucurone, D-Glucurono-6,3-lactone, Glucuronolactone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8O6
CAS Number:
Molecular Weight:
176.12
Beilstein:
83595
EC Number:
MDL number:
UNSPSC Code:
12352115
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥99%

form

powder

optical activity

[α]24/D +18.8°, c = 8 in H2O

mp

172-175 °C (lit.)

solubility

water: soluble 25 mg/mL, clear, colorless

SMILES string

O=C([C@@H]([C@@H](O1)[C@H](O)[C@H](O)C1=O)O)[H]

InChI

1S/C6H8O6/c7-1-2(8)5-3(9)4(10)6(11)12-5/h1-5,8-10H/t2-,3+,4-,5+/m0/s1

InChI key

UYUXSRADSPPKRZ-SKNVOMKLSA-N

Looking for similar products? Visit Product Comparison Guide

General description

D-(+)-Glucuronic acid γ-lactone (Glucourono-γ-lactone, Glucurone or Glycurone) is a carbohydrate derivative. It converted into L-ascorbic acid in animals and human body. Its molecule contains two five-membered rings. Its crystal structure has been studied.

Application

D-(+)-Glucuronic acid γ-lactone may be used in the following studies:
  • As starting ragent in the synthesis of 2,3,4,-tris(tert.-butyldimethysilyl) glucuronic acid trichloroethylester, required for the preparation of 1-O-acyl glucuronide of the anti-inflammatory drug ML-3000.
  • Synthesis of optically active glucopyranoses.
  • Synthesis of long-chain alkyl glucofuranosides.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Alford et al.
Amino acids, 21(2), 139-150 (2001-10-23)
The effects of Red Bull Energy Drink, which includes taurine, glucuronolactone, and caffeine amongst the ingredients, were examined over 3 studies in a total of 36 volunteers. Assessments included psychomotor performance (reaction time, concentration, memory), subjective alertness and physical endurance.
Rec. Trav. Chim., 113, 79-79 (1994)
L A Reyner et al.
Physiology & behavior, 75(3), 331-335 (2002-03-19)
Driver sleepiness is a major cause of serious road crashes. Coffee is often used as an effective countermeasure to driver sleepiness. However, the caffeine levels in coffee are variable, whereas certain proprietary "functional energy drinks" (FEDs) contain known levels of
J M Hsu et al.
The Journal of nutrition, 111(1), 141-145 (1981-01-01)
The effect of feeding 0.25% ethionine for 3-10 weeks to male and female rats on urinary and tissue ascorbate contents were studied. The concentrations of ascorbic acid in the urine, blood, liver and adrenals were significantly reduced in the rats
J A Horne et al.
Amino acids, 20(1), 83-89 (2001-04-20)
500 ml of a glucose based "energy" drink versus a control without the active ingredients (caffeine, taurine, glucuronolactone) were given double blind to 11 sleepy participants driving an interactive real-car driving simulator. Lane drifting and a secondary task (reaction time)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service