Skip to Content
Merck
All Photos(1)

Key Documents

84172

Sigma-Aldrich

Salicylaldoxime

≥98.0% (NT)

Synonym(s):

2-Hydroxybenzaldehyde oxime

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-(HO)C6H4CH=NOH
CAS Number:
Molecular Weight:
137.14
Beilstein:
1859881
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98.0% (NT)

form

crystals

mp

57-59 °C
59-61 °C (lit.)

functional group

amine

SMILES string

O\N=C\c1ccccc1O

InChI

1S/C7H7NO2/c9-7-4-2-1-3-6(7)5-8-10/h1-5,9-10H/b8-5+

InChI key

ORIHZIZPTZTNCU-VMPITWQZSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Novel uncharged triazole salicylaldoxime derivatives as potential acetylcholinesterase reactivators: This research focuses on the synthesis and evaluation of novel salicylaldoxime derivatives, aimed at reactivating acetylcholinesterase inhibited by nerve agents (MH Baghersad, A Habibi, 2023).

  • Selectivity of Salicylaldoxime and its Derivatives: This study utilizes Density Functional Theory (DFT) to examine the selectivity and effectiveness of salicylaldoxime derivatives in various chemical applications (B Tuzun, 2014).

  • Synthesis and Spectroscopic study of Pd(II)-Salicylaldoxime complexes with amine ligands: This paper presents the synthesis and characterization of new Pd(II) complexes with salicylaldoxime, exploring their potential uses in catalysis and material science (EN Al_Sabawi et al., 2021).

Other Notes

Reagent for the spectrophotometric determination of Cu(II), Fe (III), Mo(VI) and Ni(II); a review

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ross S Forgan et al.
Chemical communications (Cambridge, England), (34)(34), 4049-4051 (2008-09-02)
Attaching dialkylaminomethyl arms to commercial phenolic oxime copper extractants yields reagents which transport base metal salts very efficiently by forming neutral 1:1 or 1:2 complexes with zwitterionic forms of the ligands.
D D Ourth et al.
Developmental and comparative immunology, 6(1), 75-85 (1982-01-01)
Fresh channel catfish (Ictalurus punctatus) serum from unimmunized catfish exhibited 100% bactericidal activity against Salmonella paratyphi. Components responsible for bactericidal activity could be absorbed from the fresh catfish serum with S. paratyphi. The bactericidal system of the fresh catfish serum
F Minutolo et al.
Journal of medicinal chemistry, 44(24), 4288-4291 (2001-11-16)
The phenolic "A-ring" of natural and synthetic estrogen receptor (ER) ligands was effectively replaced by a planar six-member ring formed through an intramolecular hydrogen bond within a salicylaldoxime. Thus, oxime 1, a structural analogue of a triarylethylene estrogen, showed a
Ian A Gass et al.
Dalton transactions (Cambridge, England : 2003), (15)(15), 2043-2053 (2008-04-03)
The synthesis and magnetic properties of the compounds [HNEt(3)][Fe(2)(OMe)(Ph-sao)(2) (Ph-saoH)(2)].5MeOH (1.5MeOH), [Fe(3)O(Et-sao)(O(2)CPh)(5)(MeOH)(2)].3MeOH (2.3MeOH), [Fe(4)(Me-sao)(4)(Me-saoH)(4)] (3), [HNEt(3)](2)[Fe(6)O(2)(Me-sao)(4)(SO(4))(2)(OMe)(4)(MeOH)(2)] (4), [Fe(8)O(3)(Me-sao)(3)(tea)(teaH)(3)(O(2)CMe)(3)] (5), [Fe(8)O(3)(Et-sao)(3)(tea)(teaH)(3)(O(2)CMe)(3)] (6), and [Fe(8)O(3)(Ph-sao)(3)(tea)(teaH)(3)(O(2)CMe)(3)] (7) are reported (Me-saoH(2) is 2'-hydroxyacetophenone oxime, Et-saoH(2) is 2'-hydroxypropiophenone oxime and Ph-saoH(2) is 2-hydroxybenzophenone oxime). 1-7
Tiziano Tuccinardi et al.
Bioorganic & medicinal chemistry, 21(6), 1511-1515 (2012-09-29)
New compounds containing a novel zinc binding group (salicylaldoxime system) were identified as effective inhibitors of carbonic anhydrases (CAs). This structural motif seems to bind the catalytic zinc ion of CAs, revealing itself as a new valid alternative to the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service