Skip to Content
Merck
All Photos(1)

Key Documents

82804

Sigma-Aldrich

3-(1-Pyridinio)-1-propanesulfonate

≥97.0% (N)

Synonym(s):

1-(3-Sulfopropyl)pyridinium betain, NDSB 201, PPS

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H11NO3S
CAS Number:
Molecular Weight:
201.24
Beilstein:
1621867
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

description

zwitterionic

Assay

≥97.0% (N)

form

crystals

functional group

sulfonic acid

SMILES string

[O-]S(=O)(=O)CCC[n+]1ccccc1

InChI

1S/C8H11NO3S/c10-13(11,12)8-4-7-9-5-2-1-3-6-9/h1-3,5-6H,4,7-8H2

InChI key

REEBJQTUIJTGAL-UHFFFAOYSA-N

Application

3-(1-Pyridinio)-1-propanesulfonate can be used:
  • As a Bronsted acid ionic liquid catalyst in combination with H2SO4 and H3PO4 for the dehydration of glycerol to acrolein via semi-batch reaction technique.
  • As a catalyst in the form of sulfonated heteropoly acid salts for the synthesis of isoamyl isovalerate.
  • In the preparation of molybdenum imido alkylidene NHC catalysts, which are used in biphasic olefin metathesis.
  • To prepare water-stable SO3H-functionalized Brønsted acidic task-specific ionic liquids (TSILs).

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marek Wypych et al.
Scientific reports, 9(1), 11492-11492 (2019-08-09)
Procrastination is a self-regulatory failure in which people voluntarily but irrationally delay important tasks. Trait procrastination is estimated to affect 15-20% of the total population and leads to a significant decrease in performance, satisfaction with achievements, and quality of life.
Edgar Agaba et al.
Food and nutrition bulletin, 37(4 suppl), S142-S150 (2016-12-03)
The 2011 Uganda Nutrition Action Plan (UNAP) established 2016 maternal and child nutrition targets. However, there is a lack of routine district-level data collection to assess UNAP implementation. To use Nutrition Innovation Lab (NIL) data to inform policy makers on
Molybdenum and Tungsten Imido Alkylidene N-Heterocyclic Carbene Catalysts Bearing Cationic Ligands for Use in Biphasic Olefin Metathesis
Elser I, et al.
Chemistry?A European Journal , 23(26), 6398-6405 (2017)
Xia Li et al.
Journal of epidemiology and community health, 71(6), 521-527 (2017-01-29)
The aim of this study was to explore levels and sociodemographic correlates of physical activity (PA) over 1 week using accelerometer data. Accelerometer data was collected over 1 week from 1075 8-11-year-old children in the cross-sectional Cork Children's Lifestyle Study. Threshold values
J Wang et al.
The Journal of hospital infection, 96(2), 132-138 (2017-03-23)
Healthcare-associated infections (HCAIs) are a major health threat. There are few data about HCAI and antibiotic use in the People's Republic of China in the English literature. To describe and discuss the prevalence of HCAI and antibiotic use from the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service