Skip to Content
Merck
All Photos(1)

Key Documents

809934

Sigma-Aldrich

Ethyl methyl carbonate

battery grade, 99.9%, acid <10 ppm, H2O <10ppm

Synonym(s):

Carbonic acid ethyl methyl ester, EMC, Methyl ethyl carbonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8O3
CAS Number:
Molecular Weight:
104.10
MDL number:
UNSPSC Code:
12191602
PubChem Substance ID:
NACRES:
NA.23

grade

battery grade

Quality Level

description

Acid content: <10 ppm

Assay

99.9%

form

liquid

moisture

<10 ppm

impurities

≤10 ppm H2O
≤10 ppm acid

bp

101 °C

mp

-55 °C

density

1.01 g/cm3

application(s)

battery manufacturing

SMILES string

CCOC(=O)OC

InChI

1S/C4H8O3/c1-3-7-4(5)6-2/h3H2,1-2H3

InChI key

JBTWLSYIZRCDFO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Anhydrous ethyl methyl carbonate (EMC) is a clear, colorless liquid with a sweet, ether-like odor. It is a highly polar solvent with a moderate boiling point (101 °C) and relatively low vapor pressure, making it suitable for use in a wide range of chemical reactions and processes. EMC is an excellent solvent for many organic compounds including cellulose, ester, and amides, and it can dissolve a variety of inorganic compounds as well, including many electrolytic salts.

Application

Alkyl carbonates find applications as solvents for lithium ion battery electrolytes and the use of high quality battery grade electrolytes having extremely low water (<10 ppm) and acid (<10 ppm) contents are critical for achieving high electrochemical performance.

Legal Information

Product of MU Ionic Solutions Corp

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

71.6 °F

Flash Point(C)

22 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Huang S, et al.
Chemical Society Reviews, 44, 3079-3079 (2015)
Ding M, et al.
Journal of the Electrochemical Society, 148, A1196-A1196 (2001)

Articles

Experts discuss challenges and production processes of nickel-rich layered oxide cathode materials in energy storage systems.

Experts discuss challenges and production processes of nickel-rich layered oxide cathode materials in energy storage systems.

Experts discuss challenges and production processes of nickel-rich layered oxide cathode materials in energy storage systems.

Experts discuss challenges and production processes of nickel-rich layered oxide cathode materials in energy storage systems.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service