Skip to Content
Merck
All Photos(1)

Key Documents

807761

Sigma-Aldrich

palmitic-acid-5-hydroxy-stearic-acid

95%

Synonym(s):

5-PAHSA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H66O4
CAS Number:
Molecular Weight:
538.89
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.22

Quality Level

Assay

95%

form

powder

mp

39 °C

storage temp.

2-8°C

Related Categories

General description

Palmitic-acid-5-hydroxy-stearic-acid (5-PAHSA) is also referred to as 5-hexadecanoyloxyoctadecanoic acid (IUPAC name).

Application

5-PAHSA is composed of palmitic acid attached to stearic acid and is in the class of branched lipids referred to as fatty acid esters of hydroxy fatty acids (FAHFAs). 5-PAHSA was found at reduced levels in adipose tissue and serum of insulin-resistant mice. Levels were also found to be lower in white adipose tissue and serum of humans. Studies of oral administration of 5-PAHSA to obese, diabetic mice showed improved glucose tolerance. In characterizing the mechanism, it was found that 5-PAHSA stimulates insulin and GLP-1 secretion and also enhances glucose transport and the Glut4 transporter through activation of GPR-120.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mark M Yore et al.
Cell, 159(2), 318-332 (2014-10-11)
Increased adipose tissue lipogenesis is associated with enhanced insulin sensitivity. Mice overexpressing the Glut4 glucose transporter in adipocytes have elevated lipogenesis and increased glucose tolerance despite being obese with elevated circulating fatty acids. Lipidomic analysis of adipose tissue revealed the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service