Skip to Content
Merck
All Photos(1)

Key Documents

807745

Sigma-Aldrich

APN-COCI

Synonym(s):

4-(cyanoethynyl)benzoyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H4ClNO
Molecular Weight:
189.60
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

form

powder

Quality Level

reaction suitability

reagent type: cross-linking reagent

storage temp.

2-8°C

SMILES string

N#CC#CC1=CC=C(C(Cl)=O)C=C1

InChI

1S/C10H4ClNO/c11-10(13)9-5-3-8(4-6-9)2-1-7-12/h3-6H

InChI key

OWNJCJKKKZXTIX-UHFFFAOYSA-N

Application

APN-COCl is a bifunctional reagent appropriate for the introduction of APN function.

Other Notes

The coupling between an amine and APN-COCl should be performed in dry organic solvents (e.g. THF) in the presence of a base (e.g. TEA). The resulting conjugate can be readily coupled with thiol-containing biomolecules using the following protocol:

  • Dissolve the protein in the appropriate buffer with pH 6.5-9.0 (e.g. PBS) at 1-10 mg/mL concentration.
  • Apply the appropriate amount of the stock solution of the APN-labeled molecule (1-5 molar eq. per free cysteine residue).
  • Incubate at room temperature for 2 hours.
If necessary, purify the protein conjugate using size exclusion chromatography or ultrafiltration.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sergii Kolodych et al.
Bioconjugate chemistry, 26(2), 197-200 (2015-01-24)
Amine-to-thiol coupling is the most common route for the preparation of antibody-drug conjugates (ADC). It is usually achieved by using heterobifunctional reagents possessing an activated ester at one end and a maleimide group at the other. However, maleimide-based conjugates were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service