Skip to Content
Merck
All Photos(1)

Key Documents

805556

Sigma-Aldrich

Graphene dispersion

In water, flexo/gravure/screen printable

Synonym(s):

conductive ink, graphene ink

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C
Molecular Weight:
12.01
UNSPSC Code:
12352103
NACRES:
NA.23

product name

Graphene ink in water, flexo/gravure/screen printable

Quality Level

form

liquid

concentration

7 wt. % solids in water

sheet resistance

10 Ω/sq, 25μm thickness

particle size

500-1500 nm (exfoliated graphene flakes)

viscosity

140 cP (1000s-1)
570 cP (100s-1)

Looking for similar products? Visit Product Comparison Guide

Application

Formulated for screen printing:
  • Typical substates: glass, paper.
  • Drying Condition: 100°C for 10min.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Panagiotis G Karagiannidis et al.
ACS nano, 11(3), 2742-2755 (2017-01-20)
We report the exfoliation of graphite in aqueous solutions under high shear rate [∼ 10
Electrifying inks with 2D materials.
Felice Torrisi et al.
Nature nanotechnology, 9(10), 738-739 (2014-10-07)
Edward P Randviir et al.
Physical chemistry chemical physics : PCCP, 16(10), 4598-4611 (2014-01-25)
We report the fabrication, characterisation (SEM, Raman spectroscopy, XPS and ATR) and electrochemical implementation of novel screen-printed graphene electrodes. Electrochemical characterisation of the fabricated graphene electrodes is undertaken using an array of electroactive redox probes and biologically relevant analytes, namely:
Ethan B Secor et al.
The journal of physical chemistry letters, 6(4), 620-626 (2015-08-12)
Carbon and post-carbon nanomaterials present desirable electrical, optical, chemical, and mechanical attributes for printed electronics, offering low-cost, large-area functionality on flexible substrates. In this Perspective, recent developments in carbon nanomaterial inks are highlighted. Monodisperse semiconducting single-walled carbon nanotubes compatible with

Articles

Since its discovery little more than a decade ago,1 the two-dimensional (2D) allotrope of carbon—graphene—has been the subject of intense multidisciplinary research efforts.

Since its discovery little more than a decade ago,1 the two-dimensional (2D) allotrope of carbon—graphene—has been the subject of intense multidisciplinary research efforts.

Since its discovery little more than a decade ago,1 the two-dimensional (2D) allotrope of carbon—graphene—has been the subject of intense multidisciplinary research efforts.

Since its discovery little more than a decade ago,1 the two-dimensional (2D) allotrope of carbon—graphene—has been the subject of intense multidisciplinary research efforts.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service