Skip to Content
Merck
All Photos(2)

Key Documents

805394

Sigma-Aldrich

FK 209 Co(III) TFSI salt

Synonym(s):

tris(2-(1H-pyrazol-1-yl)-4-tert-butylpyridine)cobalt(III) tri[bis(trifluoromethane)sulfonimide]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H45CoF18O12N12S6
Molecular Weight:
1503.17
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

Quality Level

form

powder

mp

281.5 °C (exp.)

SMILES string

O=S([N-]S(=O)(C(F)(F)F)=O)(C(F)(F)F)=O.O=S([N-]S(=O)(C(F)(F)F)=O)(C(F)(F)F)=O.O=S([N-]S(=O)(C(F)(F)F)=O)(C(F)(F)F)=O.CC(C)(C)C1=CC(N2N=CC=C2)=NC=C1.CC(C)(C)C3=CC=NC(N4C=CC=N4)=C3.CC(C)(C)C5=CC=NC(N6C=CC=N6)=C5.C

General description

FK 209 Co(III) TFSI salt (FK 209) is a Co(III) based complex that is used as a p-type dopant for the hole transporting material with a power conversion efficiency in the range of 9-12%.

Application

FK 209 can be used to induce a p-doping on PCDTBT and spiro-OMeTAD, which can be used as hole transporting materials in the fabrication of perovskite solar cells (PSCs) with an efficiency of 15-16%.
Use this cobalt complexes to increase photovoltages of liquid electrolyte cells substantially or to achieve ultrahigh performance with solid state photovoltaic devices.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Acid additives enhancing the conductivity of spiro-OMeTAD toward high-efficiency and hysteresis-less planar perovskite solar cells.
Li Z, et al.
Advanced Energy Materials, 7, 15714-15723 (2016)
Efficient perovskite photovoltaic devices using chemically doped PCDTBT as a hole-transport material.
Wong-Stringer M, et al.
Journal of Material Chemistry A, 5(30), 15714-15723 (2017)
Co (III) complexes as p-dopants in solid-state dye-sensitized solar cells
Burschka J, et al.
Chemistry of Materials, 25(15), 2986-2990 (2013)
Teck Ming Koh et al.
ChemSusChem, 7(7), 1909-1914 (2014-05-23)
In this work, we report a new cobalt(III) complex, tris[2-(1H-pyrazol-1-yl)pyrimidine]cobalt(III) tris[bis(trifluoromethylsulfonyl)imide] (MY11), with deep redox potential (1.27 V vs NHE) as dopant for 2,2',7,7'-tetrakis-(N,N-di-p-methoxyphenylamine)-9,9'-spirobifluorene (spiro-OMeTAD). This dopant possesses, to the best of our knowledge, the deepest redox potential among all
Edoardo Mosconi et al.
Journal of the American Chemical Society, 134(47), 19438-19453 (2012-11-02)
We report a combined experimental and computational investigation to understand the nature of the interactions between cobalt redox mediators and TiO(2) surfaces sensitized by ruthenium and organic dyes, and their impact on the performance of the corresponding dye-sensitized solar cells

Articles

Dye-sensitized solar cells (DSCs) are 3rd generation solar cells combining the promise of high efficiency with low production costs.

Next generation solar cells have the potential to achieve conversion efficiencies beyond the Shockley-Queisser (S-Q) limit while also significantly lowering production costs.

Dr. Perini and Professor Correa-Baena discuss the latest research and effort to obtain higher performance and stability of perovskite materials.

Dr. Perini and Professor Correa-Baena discuss the latest research and effort to obtain higher performance and stability of perovskite materials.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service