Skip to Content
Merck
All Photos(1)

Key Documents

802654

Sigma-Aldrich

Aquivion® D87-25BS

greener alternative

liquid, dispersion, 25% in water, PFSA eq. wt. 870 g/mole SO3H

Synonym(s):

Aquivion® SO3H-form, Tetrafluoroethylene-perfluoro(3-oxa-4-pentenesulfonic acid) copolymer, Ethanesulfonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C4HF7O4S .C2F4)x
UNSPSC Code:
26111700
NACRES:
NA.23

Quality Level

form

dispersion
liquid

eq. wt.

870±40 g/mol (SO3H)

composition

25% in water

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

concentration

>24 % (w/w)

greener alternative category

Looking for similar products? Visit Product Comparison Guide

General description

Aquivion® PFSA dispersions are derived from a short-side-chain (SSC) copolymer of Tetrafluoroethylene and Sulfonyl Fluoride Vinyl Ether (SFVE), CF2=CF2-O-(CF2)2-SO2F, industrially produced by Syensqo Group. The resulting perfluoropolymer features functional groups in their sulfonic acid form (SO3H). Aquivion® D79-25BS is a perfluorosulfonic acid (PFSA) ionomer dispersion with an Equivalent Weight (EW) of 790 g/eq. The EW of an ionomer represents the weight of the polymer required to provide 1 mole of exchangeable protons, inversely related to the ion exchange capacity (IEC). This dispersion is chemically stabilized, with a significant proportion of polymer end groups converted to an inert trifluoromethyl (CF3) configuration, enhancing the polymer′s lifetime. Aquivion® D79-25BS contains a polymer concentration of 25 wt% in a solvent system that is over 99% water and free of ethers.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency and is intended for use as Proton Exchange Membrane (PEM) Fuel Cells. Click here for more information.

Application

The superacidity of Aquivion® PFSA resins, along with their high functional group concentration and electrochemical stability, makes them essential components in heterogeneous catalysts. Because of their excellent proton conductivity at low water uptake, these resins are used frequently in research as ionomer binders within the catalyst layers of proton exchange membranes (PEM) for fuel cells and water electrolyzers. Additionally, these resins are utilized in the fabrication of separator membranes (PEMs), either as stand-alone materials or reinforced with PTFE. They are also sometimes applied to the surfaces of specialty fluoropolymers to enhance wettability. Aquivion® D87-25BS, characterized by its short side chain and moderately high equivalent weight, is a promising ionomer for balancing catalyst activity and stability. Ionomers with higher equivalent weights, such as D87-25BS, demonstrate greater durability in long-term electrochemical stability tests. D87-25BS is commonly employed as a binder with platinum group catalysts on carbon supports for oxygen evolution or oxygen reduction reactions due to its resistance to highly aggressive chemical environments, including low pH, strong oxidants, and strong reducing agents.

Features and Benefits

  • Very high concentration of functional groups for strong proton (or Na+, K+, Li+, etc.) conductivity
  • High softening point for high temperature operation well above 80 ºC
  • Strong water retention for better fuel cell operation at low-humidity conditions for membranes or catalyst inks that are utilizing this dispersion material

Legal Information

Aquivion is a registered trademark of Syensqo Group

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Advances in electrochemical water conversion and understanding PEMFC degradation drive progress in hydrogen technologies.

Advances in electrochemical water conversion and understanding PEMFC degradation drive progress in hydrogen technologies.

Advances in electrochemical water conversion and understanding PEMFC degradation drive progress in hydrogen technologies.

Advances in electrochemical water conversion and understanding PEMFC degradation drive progress in hydrogen technologies.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service