Skip to Content
Merck
All Photos(4)

Key Documents

798673

Sigma-Aldrich

Fumaric acid

anhydrous, free-flowing, Redi-Dri, ≥99%

Synonym(s):

(2E)-2-Butenedioic acid, trans-Butenedioic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOOCCH=CHCOOH
CAS Number:
Molecular Weight:
116.07
Beilstein:
605763
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

grade

anhydrous

Quality Level

vapor pressure

1.7 mmHg ( 165 °C)

product line

Redi-Dri

Assay

≥99%

form

powder

autoignition temp.

1364 °F

quality

free-flowing

expl. lim.

40 %

mp

298-300 °C (subl.) (lit.)

functional group

carboxylic acid

SMILES string

OC(=O)\C=C\C(O)=O

InChI

1S/C4H4O4/c5-3(6)1-2-4(7)8/h1-2H,(H,5,6)(H,7,8)/b2-1+

InChI key

VZCYOOQTPOCHFL-OWOJBTEDSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

523.4 °F

Flash Point(C)

273 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service