Skip to Content
Merck
All Photos(4)

Key Documents

798630

Sigma-Aldrich

Sodium formate

anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%

Synonym(s):

Formic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCOONa
CAS Number:
Molecular Weight:
68.01
Beilstein:
3595134
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥99%
grade:
ACS reagent
anhydrous
form:
powder

grade

ACS reagent
anhydrous

Quality Level

product line

Redi-Dri

Assay

≥99%

form

powder

quality

free-flowing

impurities

≤0.005% insolubles

mp

259-262 °C (lit.)

anion traces

chloride (Cl-): ≤0.001%
sulfate (SO42-): ≤0.001%

cation traces

Ca: ≤0.005%
Fe: ≤5 ppm
heavy metals (as Pb): ≤5 ppm

SMILES string

[Na+].[O-]C=O

InChI

1S/CH2O2.Na/c2-1-3;/h1H,(H,2,3);/q;+1/p-1

InChI key

HLBBKKJFGFRGMU-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Priscilla Rocío-Bautista et al.
Talanta, 215, 120910-120910 (2020-04-22)
Chemical vapor deposition of MOFs (MOF-CVD) has been used to coat solid-phase microextraction (SPME) fibers with ZIF-8, by exposing ZnO layers to the linker vapor (2-methylimidazole). This ZIF-8 coating has been used as a seed layer in a following solvothermal
Yaeseong Hong et al.
Biotechnology and bioengineering, 118(3), 1366-1380 (2020-12-18)
Autotrophic or mixotrophic use of one-carbon (C1) compounds is gaining importance for sustainable bioproduction. In an effort to integrate the reductive glycine pathway (rGP) as a highly promising pathway for the assimilation of CO2 and formate, genes coding for glycine
Enrique Javier Carrasco-Correa et al.
Talanta, 164, 348-354 (2017-01-22)
A series of metal organic frameworks-derived nanoporous carbons originating from zeolitic imidazolate framework-8 (ZIF-8) crystals as precursors have been prepared via varying the preparation conditions. The ZIF-8-derived carbons were subsequently admixed in the methacrylate monomers containing polymerization mixtures and polymerized
Ana Belén Esteban García et al.
Environmental science and pollution research international, 28(7), 8576-8584 (2020-10-17)
Laundry sector consumes a huge amount of water which is usually discharged as wastewater instead of being reused. The application of biological treatment of laundry wastewater coupled with post-treatment utilizing advanced oxidation processes creates a possibility to recycle water to
Samuel Berens et al.
Physical chemistry chemical physics : PCCP, 20(37), 23967-23975 (2018-09-14)
Pulsed field gradient (PFG) NMR was used in combination with single crystal IR microscopy (IRM) to study diffusion of ethane inside crystals of a mixed linker zeolitic imidazolate framework (ZIF) of the type ZIF-7-8 under comparable experimental conditions. These crystals

Articles

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service