Skip to Content
Merck
All Photos(2)

Key Documents

797170

Sigma-Aldrich

Neo-decanoic acid

Synonym(s):

7,7-Dimethyloctanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H20O2
CAS Number:
Molecular Weight:
172.26
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

description

Fp: >230°F

form

liquid

refractive index

n20/D 1.439

density

0.9086 g/mL at 25 °C

functional group

carboxylic acid

SMILES string

OC(=O)CCCCCC(C)(C)C

InChI

1S/C10H20O2/c1-10(2,3)8-6-4-5-7-9(11)12/h4-8H2,1-3H3,(H,11,12)

InChI key

YPIFGDQKSSMYHQ-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service