Skip to Content
Merck
All Photos(1)

Key Documents

79522

Sigma-Aldrich

tert-Butyl P,P-dimethylphosphonoacetate

≥97.0% (GC)

Synonym(s):

tert-Butyl ((oxo)dimethoxyphosphino)acetate, tert-Butyl (dimethoxyphosphinyl)acetate, Dimethyl (tert-butoxycarbonyl)methylphosphonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)2P(O)CH2COOC(CH3)3
CAS Number:
Molecular Weight:
224.19
Beilstein:
4861886
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥97.0% (GC)

form

liquid

reaction suitability

reaction type: C-C Bond Formation

refractive index

n20/D 1.434

bp

86-87 °C/0.02 mmHg (lit.)

density

1.131 g/mL at 20 °C (lit.)

SMILES string

COP(=O)(CC(=O)OC(C)(C)C)OC

InChI

1S/C8H17O5P/c1-8(2,3)13-7(9)6-14(10,11-4)12-5/h6H2,1-5H3

InChI key

SAZYDWOWLRDDRQ-UHFFFAOYSA-N

Application

Reactant for:
  • Preparation of phosphonate terminated PPH dendrimers as anti-HIV-1 agents
  • Preparation of monodehydro diketopiperazines from ketoacyl amino acid amides via acid-catalyzed cyclization
  • Preparation of α,β-unsaturated esters for use in the guanidine-catalyzed oxa-Michael addition
  • Preparation of myxopyronin B and desmethyl myxopyronin B analogs, with antibacterial activity and inhibitory activity against bacterial RNA polymerase
  • Allylation and subsequent ring-closing metathesis in presence of Grubbs′ catalyst or intramolecular rhodium-catalyzed cyclopropanation reactions

Other Notes

Horner-Wittig reagent giving preferentially (E)-α,β-unsat. esters which are mildly saponified with acid. Synthesis of derivatives by reductive alkylation

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A novel synthesis of (+/-)-vermiculine.
Y Fukuyama et al.
Journal of the American Chemical Society, 99(2), 646-647 (1977-01-19)
W.J. Guilford et al.
Journal of the American Chemical Society, 109, 5013-5013 (1987)
G.W.J. Fleet et al.
Journal of the Chemical Society. Perkin Transactions 1, 905-905 (1984)
C. Kuroda et al.
The Journal of Organic Chemistry, 51, 956-956 (1986)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service