Skip to Content
Merck
All Photos(3)

Key Documents

794325

Sigma-Aldrich

Basolite® Z377

Synonym(s):

MOF 177

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C54H30O13Zn4
CAS Number:
Molecular Weight:
1148.37
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.23

description

Composition: Carbon (C) 55 g/100g, Zinc (Zn) 22 g/100g ±10%
Surface area: Lanmuir Suface area = 5000-5200 m2/g, Activation: T = 200°C, p <0.1mbar, t = 16h

form

powder

surface area

3800-4000 m2/g , Activation: T = 200°C, p <0.1mbar, t = 16

SMILES string

[O-]C(C1=CC=C(C2=CC(C3=CC=C(C([O-])=O)C=C3)=CC(C4=CC=C(C(O[Zn+])O[Zn+]O[Zn+]OC(C5=CC=C(C6=CC(C7=CC=C(C([O-])=O)C=C7)=CC(C8=CC=C(C([O-])=O)C=C8)=C6)C=C5)O[Zn+])C=C4)=C2)C=C1)=O

InChI

1S/2C27H18O6.O.4Zn/c2*28-25(29)19-7-1-16(2-8-19)22-13-23(17-3-9-20(10-4-17)26(30)31)15-24(14-22)18-5-11-21(12-6-18)27(32)33;;;;;/h2*1-15,25H,(H,30,31)(H,32,33);;;;;/q2*-2;;4*+2/p-4

InChI key

CLMVSZQXLQAGTL-UHFFFAOYSA-J

Application

Basolite® Z377, also known as MOF 177, is a commonly used metal organic framework for hydrogen absorption.

Legal Information

Basolite is a registered trademark of BASF SE

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hydrogen physisorption in high SSA microporous materials - A comparison between AX-2133 and MOF-177 at cryogenic conditions
Schlichtenmayer, et al.
International Journal of Hydrogen Energy, 36(1), 586-591 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service