Skip to Content
Merck
All Photos(1)

Key Documents

79432

Sigma-Aldrich

Phosphazene base P1-t-Bu-tris(tetramethylene)

≥97.0% (NT)

Synonym(s):

tert-Butylimino-tri(pyrrolidino)phosphorane, BTPP, BTTP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H33N4P
CAS Number:
Molecular Weight:
312.43
Beilstein:
9100965
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥97.0% (NT)

form

liquid

refractive index

n20/D 1.509

bp

119-122 °C/0.1 mmHg (lit.)

mp

−24 °C (lit.)

density

1.022 g/mL at 20 °C (lit.)

functional group

amine

storage temp.

2-8°C

SMILES string

CC(C)(C)N=P(N1CCCC1)(N2CCCC2)N3CCCC3

InChI

1S/C16H33N4P/c1-16(2,3)17-21(18-10-4-5-11-18,19-12-6-7-13-19)20-14-8-9-15-20/h4-15H2,1-3H3

InChI key

PVNUIRUAPVSSOK-UHFFFAOYSA-N

Other Notes

This phosphazene base is 1.4 pK units more basic than the hexamethyl analogue (Cat. No. 79408)citationcitation; Compare also with the analogue BEMP (Cat. No. 20025)
This phosphazene base is 1.4 pK units more basic than the hexamethyl analogue (Cat. No. 79408); Compare also with the analogue BEMP (Cat. No. 20025)

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemische Berichte, 127, 2435-2435 (1994)
R. Schwesinger et al
Angewandte Chemie (International Edition in English), 105, 1420-1420 (1993)

Articles

Phosphazene base reagents are available as monomeric (P1 and BEMP), dimeric (P2), and tetrameric (P4) bases with different side chains to control their sterical hindrance.

Phosphazene base reagents are available as monomeric (P1 and BEMP), dimeric (P2), and tetrameric (P4) bases with different side chains to control their sterical hindrance.

Phosphazene base reagents are available as monomeric (P1 and BEMP), dimeric (P2), and tetrameric (P4) bases with different side chains to control their sterical hindrance.

Phosphazene base reagents are available as monomeric (P1 and BEMP), dimeric (P2), and tetrameric (P4) bases with different side chains to control their sterical hindrance.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service