Skip to Content
Merck
All Photos(1)

Key Documents

779431

Sigma-Aldrich

γ-Cyclodextrin

produced by Wacker Chemie AG, Burghausen, Germany, ≥90.0% cyclodextrin basis (HPLC)

Synonym(s):

Cavamax® W8, gamma-Cyclodextrin, Cyclomaltooctaose, Cyclooctaamylose, Schardinger γ-Dextrin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C48H80O40
CAS Number:
Molecular Weight:
1297.12
Beilstein:
78740
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.22

grade

produced by Wacker Chemie AG, Burghausen, Germany

Quality Level

Assay

≥90.0% cyclodextrin basis (HPLC)

form

powder

loss

≤11.0% loss on drying

SMILES string

[H][C@]1(O[C@H](COC(C)=O)[C@@H](OC(C)=O)[C@H](OC(C)=O)[C@H]1OC(C)=O)O[C@@H]2[C@@H](COC(C)=O)O[C@H](OC(C)=O)[C@H](OC(C)=O)[C@H]2OC(C)=O

InChI

1S/C48H80O40/c49-1-9-33-17(57)25(65)41(73-9)82-34-10(2-50)75-43(27(67)19(34)59)84-36-12(4-52)77-45(29(69)21(36)61)86-38-14(6-54)79-47(31(71)23(38)63)88-40-16(8-56)80-48(32(72)24(40)64)87-39-15(7-55)78-46(30(70)22(39)62)85-37-13(5-53)76-44(28(68)20(37)60)83-35-11(3-51)74-42(81-33)26(66)18(35)58/h9-72H,1-8H2/t9-,10-,11-,12-,13-,14-,15-,16-,17-,18-,19-,20-,21-,22-,23-,24-,25-,26-,27-,28-,29-,30-,31-,32-,33-,34-,35-,36-,37-,38-,39-,40-,41-,42-,43-,44-,45-,46-,47-,48-/m1/s1

InChI key

GDSRMADSINPKSL-HSEONFRVSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Gamma-cyclodextrin (γ-CD) is a cyclic α-1,4-glucan linked oligosaccharide that is widely used in the food and pharmaceutical industries. In drug delivery, γ-CD can be used as a complexing agent because it forms an inclusion complex with a drug molecule. The complex of cyclodextrin increases the aqueous solubility and dissolution rate of the drug, which is useful for the delivery of the medical agent to a biological system.

Application

γ-Cyclodextrin can be used as a precursor for the synthesis of:
  • Octakis-(2,3-di-O-methyl-6-O-carboxymethyl)-γ-cyclodextrin sodium salt (ODMCM). ODMCM is used in capillary electrophoresis as a chiral resolving agent.
  • Water-soluble cyclodextrin thioethers derivatives for the transportation of hydrophobic drugs.

Legal Information

Cavasol is a registered trademark of Wacker Chemie AG

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of uniform cyclodextrin thioethers to transport hydrophobic drugs
Becker LF, et al.
Beilstein Journal of Organic Chemistry, 10(1), 2920-2927 (2014)
Safety assessment of γ-cyclodextrin
Munro IC, et al.
Regulatory Toxicology and Pharmacology, 39, 3-13 (2004)
Single-isomer carboxymethyl-γ-cyclodextrin as chiral resolving agent for capillary electrophoresis
Benkovics G, et al.
Journal of Chromatography A, 1467(2), 445-453 (2016)
Phatsawee Jansook et al.
Journal of pharmacy & pharmaceutical sciences : a publication of the Canadian Society for Pharmaceutical Sciences, Societe canadienne des sciences pharmaceutiques, 13(3), 336-350 (2010-11-26)
Study the complexation of dexamethasone in combinations of γ-cyclodextrin (γCD) and 2-hydroxypropyl-γ-cyclodextrin (HPγCD) with emphasis on solid characterization and development of aqueous dexamethasone eye drop suspension for drug delivery through sclera. Dexamethasone/cyclodextrin (dexamethasone/CD) solid complex systems were prepared and characterized
B-G Kim et al.
British journal of pharmacology, 164(6), 1698-1710 (2011-05-19)
Although 3α-hydroxy, 5α-reduced pregnane steroids, such as allopregnanolone (AlloP) and tetrahydrodeoxycorticosterone, are endogenous positive modulators of postsynaptic GABA(A) receptors, the functional roles of endogenous neurosteroids in synaptic transmission are still largely unknown. In this study, the effect of AlloP on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service