Skip to Content
Merck
All Photos(1)

Key Documents

776661

Sigma-Aldrich

Gold nanorods

10 nm diameter, λmax, 980 nm, dispersion in H2O

Synonym(s):

Au nanorod, Gold nanorod, Au nanorods

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

form

colloidal suspension
dispersion in H2O
nanorod

contains

CTAB as stabilizer

concentration

>30 μg/mL in H2O

diam. × L

10 nm × 59 nm (±10%)

diameter

10 nm

density

1.00 g/mL at 25 °C

λmax

980 nm

Mw/Mn

<10 (CV, monodispersity)

storage temp.

2-8°C

SMILES string

[Au]

InChI

1S/Au

InChI key

PCHJSUWPFVWCPO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

High aspect ratio nanorods allow the extension of the absorption peak into the IR; making them ideal for diagnostic applications in biological systems.

Publication link:
Gold Nanostructures: Properties and Applications

Physical properties

Do not freeze, quality lost if frozen even once. Please keep at approximately 4 °C

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Prashant K Jain et al.
The journal of physical chemistry. B, 110(14), 7238-7248 (2006-04-08)
The selection of nanoparticles for achieving efficient contrast for biological and cell imaging applications, as well as for photothermal therapeutic applications, is based on the optical properties of the nanoparticles. We use Mie theory and discrete dipole approximation method to

Articles

Gold nanostructures such as nanorods, nanowires and microgold have found applications in exciting fields such as biomedical engineering, catalysis and diagnostics.

Gold nanostructures such as nanorods, nanowires and microgold have found applications in exciting fields such as biomedical engineering, catalysis and diagnostics.

Gold nanostructures such as nanorods, nanowires and microgold have found applications in exciting fields such as biomedical engineering, catalysis and diagnostics.

Gold nanostructures such as nanorods, nanowires and microgold have found applications in exciting fields such as biomedical engineering, catalysis and diagnostics.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service