Skip to Content
Merck
All Photos(7)

Key Documents

776033

Sigma-Aldrich

SPGS-550-M

greener alternative

2% w/w in H2O

Synonym(s):

Nok

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C58H104O16
Molecular Weight:
1057.44
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

form

liquid

Quality Level

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

concentration

2% w/w in H2O

refractive index

n20/D 1.336

density

1.005 g/mL at 25 °C

greener alternative category

storage temp.

2-8°C

SMILES string

CC(C)C(CC)CCC(C)C1CCC2C3CC=C4CC(OC(CCC(OCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOC)=O)=O)CCC4(C)C3CCC21C

InChI

1S/C58H104O16/c1-8-48(46(2)3)10-9-47(4)52-13-14-53-51-12-11-49-45-50(17-19-57(49,5)54(51)18-20-58(52,53)6)74-56(60)16-15-55(59)73-44-43-72-42-41-71-40-39-70-38-37-69-36-35-68-34-33-67-32-31-66-30-29-65-28-27-64-26-25-63-24-23-62-22-21-61-7/h11,46-48,50-54H,8-10,12-45H2,1-7H3

InChI key

YTKPTELRRARCNX-UHFFFAOYSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

Third Generation Designer Surfactant, in collaboration with the Lipshutz Group, is the most cost-effective surfactant to this point. "Nok" provides comparable yields for several cross-coupling reactions to first generation PTS (698717) and second generation TPGS (733857) at room temperature.

"Nok":A Phytosterol-Based Amphiphile Enabling Transition-Metal-Catalyzed Couplings in Water at Room Temperature

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nihar R Desai et al.
PloS one, 15(1), e0226844-e0226844 (2020-01-08)
Renin-angiotensin-aldosterone system inhibitor (RAASi) therapy has been shown to improve outcomes among patients with congestive heart failure, diabetes, or renal dysfunction. These patients are also at risk for the development of hyperkalemia (HK), often leading to down-titration and/or discontinuation of
Carmel Martin et al.
Journal of evaluation in clinical practice, 27(2), 228-235 (2020-08-29)
HealthLinks: Chronic Care is a state-wide public hospital initiative designed to improve care for cohorts at-risk of potentially preventable hospitalizations at no extra cost. MonashWatch (MW) is an hospital outreach service designed to optimize admissions in an at-risk cohort. Telehealth

Articles

Micellular catalysis has provided the ability to carry out several commonly used transformations used in the synthetic community to be carried out in water.

Surfactants for efficient reactions in water as a green chemistry alternative.

Surfactants for efficient reactions in water as a green chemistry alternative.

Surfactants for efficient reactions in water as a green chemistry alternative.

See All

Related Content

Prof. Bruce Lipshutz and co-workers have developed designer surfactants to allow several classes of transformations (e.g. Suzuki-Miyaura, Olefin Metathesis, 1,4-Addition to Enones, etc.) to be performed in water.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service