Skip to Content
Merck
All Photos(1)

Key Documents

775797

Sigma-Aldrich

Isopropylmagnesium bromide solution

0.75 M in THF

Synonym(s):

1-Methylethylmagnesium bromide, 2-Propylmagnesium bromide, iso-Propylmagnesium bromide, Bromoisopropylmagnesium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2MgBr
CAS Number:
Molecular Weight:
147.30
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

form

solution

reaction suitability

reaction type: Grignard Reaction

concentration

0.75 M in THF

density

0.982 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

CC(C)[Mg]Br

InChI

1S/C3H7.BrH.Mg/c1-3-2;;/h3H,1-2H3;1H;/q;;+1/p-1

InChI key

UCJIVFREPMUZDC-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

  • Preparation of isopropylmagnesium bromide solutions: In this study, various concentrations of isopropylmagnesium bromide solutions were prepared, detailing their stability and reactivity for use in palladium-catalyzed cross-coupling reactions and other synthetic applications (Vulović et al., 2017).
  • Reductive coupling of amides: This research discusses the reductive functionalization of tertiary amides using isopropylmagnesium bromide, providing a novel method for amine synthesis, which is crucial for pharmaceutical chemistry (Xie and Dixon, 2017).
  • Synthesis of 1, 2, 3-triazoles: The study explores the use of isopropylmagnesium bromide in the synthesis of 1, 2, 3-triazoles through Grignard reactions, highlighting its role in protecting azides during the reaction process (Namioka et al., 2023).
  • Disposable cartridge concept for Grignard reagents: This paper introduces a novel disposable cartridge for the on-demand synthesis of Grignard reagents, including isopropylmagnesium bromide, enhancing the safety and scalability of Grignard reactions (Berton et al., 2020).

Packaging

The 25 mL Sure/Seal bottle is recommended as a single-use bottle. Repeated punctures will likely result in decreased performance of product.

Legal Information

Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3 - Water-react 1

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 1

Flash Point(F)

-22.0 °F - closed cup

Flash Point(C)

-30 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service