isotopic purity
98 atom % D
Assay
98%
form
liquid
mass shift
M+21
SMILES string
[2H]C([2H])(C([2H])(C([2H])(C([2H])(C([2H])(C([2H])(C([2H])(C([2H])(C([2H])(C([2H])(O)[2H])[2H])[2H])[2H])[2H])[2H])[2H])[2H])[2H])[2H]
InChI
1S/C10H22O/c1-2-3-4-5-6-7-8-9-10-11/h11H,2-10H2,1H3/i1D3,2D2,3D2,4D2,5D2,6D2,7D2,8D2,9D2,10D2
InChI key
MWKFXSUHUHTGQN-SLBGAMDCSA-N
Related Categories
Packaging
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2 - Eye Irrit. 2
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
179.6 °F - closed cup
Flash Point(C)
82 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service