Skip to Content
Merck
All Photos(1)

Key Documents

771619

Sigma-Aldrich

Manganese(II) fluoride

≥99.95% trace metals basis

Synonym(s):

Manganese bifluoride, Manganese difluoride, Manganous fluoride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MnF2
CAS Number:
Molecular Weight:
92.93
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

grade

anhydrous

Quality Level

Assay

≥99.95% trace metals basis

form

powder

impurities

≤500.0 ppm Trace Metal Analysis

mp

856 °C (lit.)

density

3.98 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

F[Mn]F

InChI

1S/2FH.Mn/h2*1H;/q;;+2/p-2

InChI key

CTNMMTCXUUFYAP-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Anhydrous Manganese(II) Fluoride is an interesting antiferromagnetic material as well as a precursor for lithium ion battery materials.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S P Bayrakci et al.
Science (New York, N.Y.), 312(5782), 1926-1929 (2006-07-01)
We used a neutron spin-echo method with microelectron-volt resolution to determine the lifetimes of spin waves in the prototypical antiferromagnet MnF2 over the entire Brillouin zone. A theory based on the interaction of spin waves (magnons) with longitudinal spin fluctuations
Gocheva; I.;
Journal of Power Sources, 187(1), 247-252 (2009)

Articles

Lithium-ion batteries' characteristics make them popular for electricity storage due to portability, rechargeability, and low cost.

Lithium-ion batteries' characteristics make them popular for electricity storage due to portability, rechargeability, and low cost.

Lithium-ion batteries' characteristics make them popular for electricity storage due to portability, rechargeability, and low cost.

Lithium-ion batteries' characteristics make them popular for electricity storage due to portability, rechargeability, and low cost.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service