Skip to Content
Merck
All Photos(1)

Key Documents

764760

Sigma-Aldrich

Poly(ethylene glycol) methyl ether-block-poly(lactide-co-glycolide)

PEG average Mn 2,000, PLGA average Mn 11,500

Synonym(s):

PEG-PLGA, Polyethylene glycol, mPEG-b-PLGA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H[(C3H4O2)x(C2H2O2)y]mO[C2H4O]nCH3
UNSPSC Code:
12162002
NACRES:
NA.23

form

pellets

Quality Level

feed ratio

lactide:glycolide 50:50

mol wt

PEG average Mn 2,000
PLGA average Mn 11,500
average Mn 13,500 (total)

degradation timeframe

1-4 weeks

transition temp

Tm 298-303 °C
Tg 40 °C (PDLLA block)
Tg 6 °C (PEG block)

PDI

≤2.0

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Amphiphilic block copolymers (AmBC) are made up of two chemically different homopolymer blocks. One of the block is hydrophilic and the other one is hydrophobic. These macromolecules have the properties to self-assemble when dissolved in an aqueous media. PEG-PLGA is one the most commonly used biodegradable amphiphilic block copolymers for drug delivery applications. PEG is the hydrophilic part and PLGA is the hydrophobic part.

Application

Used in the synthesis of targeted nanoparticles which are used for differential delivery and controlled release of drugs.

Features and Benefits

  • Good biocompatibility, low immunogenicity and good degradability.
  • Properties can be easily modulated by changing the block copolymer segment sizes to suit a particular application.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Che-Ming Jack Hu et al.
International journal of nanomedicine, 13, 8579-8593 (2018-12-28)
Influenza virus infections are a major public health concern worldwide. Conventional treatments against the disease are designed to target viral proteins. However, the emergence of viral variants carrying drug-resistant mutations can outpace the development of pathogen-targeting antivirals. Diphyllin and bafilomycin
Frank Gu et al.
Proceedings of the National Academy of Sciences of the United States of America, 105(7), 2586-2591 (2008-02-15)
There has been progressively heightened interest in the development of targeted nanoparticles (NPs) for differential delivery and controlled release of drugs. Despite nearly three decades of research, approaches to reproducibly formulate targeted NPs with the optimal biophysicochemical properties have remained
Thermosensitive self-assembling block copolymers as drug delivery systems.
Bonacucina, G., Cespi, M., Mencarelli, G., Giorgioni, G., & Palmieri, G. F.
Polymer, 3(2), 779-811 (2011)
PLGA-PEG Encapsulated sitamaquine nanoparticles drug delivery system against Leishmania donovani
Kumara, R., Sahoo, G. C., Pandeya, K., Dasa, V. N. R., Yousuf, M., Ansaria, S. R., & Dasa, P.
Journal of Scientific and Innovative Research, 3(1), 85-90 (2014)

Articles

Micelle formation addresses low solubility in IV drug delivery, overcoming clinical limitations.

Micelle formation addresses low solubility in IV drug delivery, overcoming clinical limitations.

Micelle formation addresses low solubility in IV drug delivery, overcoming clinical limitations.

Micelle formation addresses low solubility in IV drug delivery, overcoming clinical limitations.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service