Skip to Content
Merck
All Photos(1)

Key Documents

76172

Sigma-Aldrich

O-(2-Aminoethyl)-O′-(2-azidoethyl)pentaethylene glycol

≥90% (oligomer purity)

Synonym(s):

Azido-PEG-amine (n=6)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H30N4O6
CAS Number:
Molecular Weight:
350.41
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥90% (oligomer purity)

form

liquid

reaction suitability

reaction type: click chemistry
reagent type: cross-linking reagent

functional group

amine
azide

storage temp.

2-8°C

SMILES string

NCCOCCOCCOCCOCCOCCOCCN=[N+]=[N-]

InChI

1S/C14H30N4O6/c15-1-3-19-5-7-21-9-11-23-13-14-24-12-10-22-8-6-20-4-2-17-18-16/h1-15H2

InChI key

VCQSTKKJKNUQBI-UHFFFAOYSA-N

Application

O-(2-Aminoethyl)-O′-(2-azidoethyl)pentaethylene glycol can be used:
  • As a reactant for the synthesis of thin films of azide functionalized poly(L-glutamic acid) for drug delivery.
  • For the synthesis of curcumin monoazide derivatives for fabricating biologically active curcumin conjugates.
  • To modify the surface of TiO2 nanoparticles to attach DNA strands for photocatalytic reduction of CO2.
  • For the conjugation of poly(ethylene glycol) (PEG) with toll-like receptor 7 (TLR7) to improve its pharmaceutical properties.

O-(2-Aminoethyl)-O′-(2-azidoethyl)pentaethylene glycol has been used in the synthesis of Alexa568-azide, a fluorescent azide that can undergo click staining reaction with 5-ethynyluridine-labeled cellular RNA.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of monofunctional curcumin derivatives, clicked curcumin dimer, and a PAMAM dendrimer curcumin conjugate for therapeutic applications.
Shi W, et al.
Organic Letters, 9(26), 5461-5464 (2007)
Exploring RNA transcription and turnover in vivo by using click chemistry.
Jao CY & Salic A.
Proceedings of the National Academy of Sciences of the USA, 105(41), 15779-15784 (2008)
DNA for Assembly and Charge Transport Photocatalytic Reduction of CO2.
Ma K, et al.
Advanced Sustainable Systems, 2(4), 1700156-1700156 (2018)
Low-fouling, biofunctionalized, and biodegradable click capsules.
Ochs CJ, et al.
Biomacromolecules, 9(12), 3389-3396 (2008)
Metabolic labeling and direct imaging of choline phospholipids in vivo.
Jao CY, et al.
Proceedings of the National Academy of Sciences of the USA, 106(36), 15332-15337 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service