Skip to Content
Merck
All Photos(1)

Key Documents

761281

Sigma-Aldrich

Chloro[(1,3,5,7-tetramethyl-5-phenyl-2,4,8-trioxa-6-phosphaadamantane)-2-(2-aminobiphenyl)]palladium(II)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H31ClNO3PPd
CAS Number:
Molecular Weight:
602.40
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

form

solid

Quality Level

mp

202-208 °C (decomposition)

SMILES string

Nc1ccccc1-c2ccccc2[Pd]Cl.CC34CC5(C)OC(C)(CC(C)(O3)P5c6ccccc6)O4

InChI

1S/C16H21O3P.C12H10N.ClH.Pd/c1-13-10-15(3)19-14(2,17-13)11-16(4,18-13)20(15)12-8-6-5-7-9-12;13-12-9-5-4-8-11(12)10-6-2-1-3-7-10;;/h5-9H,10-11H2,1-4H3;1-6,8-9H,13H2;1H;/q;;;+1/p-1

InChI key

RMRMUXKLTCTPCF-UHFFFAOYSA-M

Related Categories

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service