Skip to Content
Merck
All Photos(1)

Key Documents

760498

Sigma-Aldrich

Nitric acid-18O3

95 atom % 18O, 65 wt. % in H218O

Synonym(s):

18O Labeled nitric acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HN18O3 solution
CAS Number:
UNSPSC Code:
12352116
PubChem Substance ID:

isotopic purity

95 atom % 18O

form

liquid

concentration

65 wt. % in H218O

mass shift

M+6

SMILES string

[18OH][N+]([18O-])=[18O]

InChI

1S/HNO3/c2-1(3)4/h(H,2,3,4)/i2+2,3+2,4+2

InChI key

GRYLNZFGIOXLOG-PGVNZGOUSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Eye Dam. 1 - Skin Corr. 1A

Supplementary Hazards

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service