Skip to Content
Merck
All Photos(1)

Key Documents

756822

Sigma-Aldrich

Deuterium oxide

filtered, 99.8 atom % D

Synonym(s):

Deuterated water, Heavy water, Water-d2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
D2O
CAS Number:
Molecular Weight:
20.03
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.12

Quality Level

isotopic purity

99.8 atom % D

form

liquid

technique(s)

HPLC: suitable
bio NMR: suitable
protein expression: suitable

bp

101.4 °C (lit.)

mp

3.8 °C (lit.)

density

1.107 g/mL at 25 °C

application(s)

pharmaceutical

storage temp.

room temp

SMILES string

[2H]O[2H]

InChI

1S/H2O/h1H2/i/hD2

InChI key

XLYOFNOQVPJJNP-ZSJDYOACSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Deuterium oxide (D2O) is also known as heavy water. It is the deuterated form of water, as both the protons in H2O have been replaced with a deuterium isotope.

Application

D2O can be used as a solvent in:      
  • High-resolution NMR (Nuclear magnetic resonance)      
  • FTIR (Fourier-transform infrared)      
  • Bio NMR studies      
  • Polarized visible Raman spectroscopic studies      
  • Protein expression

D2O is also used to prepare deuterium-labeled compounds in various fields such as pharmaceutical, bioanalytical, biological, and environmental chemistry via H-D exchange reactions to study drug metabolism mechanisms, reaction mechanisms, and the structure of biomolecules, and residual agrochemicals in foods using suitable analytical methods.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

R Schweitzer-Stenner et al.
Journal of the American Chemical Society, 123(39), 9628-9633 (2001-09-27)
We have measured the polarized visible Raman and FTIR spectra of trialanine and triglycine in D(2)O at acid, neutral, and alkaline pD. From the Raman spectra we obtained the isotropic and the anisotropic scattering. A self-consistent spectral analysis of the
Raphael Böhm et al.
Structure (London, England : 1993), 28(2), 206-214 (2019-12-22)
The voltage-dependent anion channel (VDAC) forms the primary diffusion pore of the outer mitochondrial membrane. In its apo form, VDAC adopts an open conformation with high conductance. States of lower conductance can be induced by ligand binding or the application
Liset Westera et al.
Blood, 122(13), 2205-2212 (2013-08-16)
Quantitative knowledge of the turnover of different leukocyte populations is a key to our understanding of immune function in health and disease. Much progress has been made thanks to the introduction of stable isotope labeling, the state-of-the-art technique for in
Lars Holm et al.
American journal of physiology. Endocrinology and metabolism, 304(8), E895-E907 (2013-02-21)
A method to determine the rate of protein breakdown in individual proteins was developed and tested in rats and confirmed in humans, using administration of deuterium oxide and incorporation of the deuterium into alanine that was subsequently incorporated into body
Takhar Kasumov et al.
American journal of physiology. Heart and circulatory physiology, 304(9), H1201-H1214 (2013-03-05)
Traditional proteomics provides static assessment of protein content, but not synthetic rates. Recently, proteome dynamics with heavy water ((2)H2O) was introduced, where (2)H labels amino acids that are incorporated into proteins, and the synthesis rate of individual proteins is calculated

Related Content

Protein structure analysis aids in identifying disease biomarkers and drug targets crucial for therapeutic treatments.

Protein structure analysis aids in identifying disease biomarkers and drug targets crucial for therapeutic treatments.

Protein structure analysis aids in identifying disease biomarkers and drug targets crucial for therapeutic treatments.

Protein structure analysis aids in identifying disease biomarkers and drug targets crucial for therapeutic treatments.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service