756245
2,4,6,8-Tetramethyl-2,4,6,8-tetrakis(propyl glycidyl ether)cyclotetrasiloxane
90%
Synonym(s):
CS-697, Polyglycidyl ether cyclosiloxane, Tetrakis(propyl glycidyl ether)cyclomethylsiloxane
About This Item
Recommended Products
Assay
90%
form
viscous liquid
eq. wt.
175-190 g/eq Epoxide
viscosity
125-250 cP
density
1.132 g/mL at 25 °C
storage temp.
2-8°C
SMILES string
C[Si]2(CCCOCC1CO1)O[Si](C)(CCCOCC3CO3)O[Si](C)(CCCOCC4CO4)O[Si](C)(CCCOCC5CO5)O2
InChI
1S/C28H56O12Si4/c1-41(13-5-9-29-17-25-21-33-25)37-42(2,14-6-10-30-18-26-22-34-26)39-44(4,16-8-12-32-20-28-24-36-28)40-43(3,38-41)15-7-11-31-19-27-23-35-27/h25-28H,5-24H2,1-4H3
InChI key
BLZDTVROPKIJLZ-UHFFFAOYSA-N
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
392.0 °F
Flash Point(C)
200 °C
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service