755206
1,3-Difluoroacetone
97%
Synonym(s):
1,3-Difluoro-2-propanone
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Assay
97%
form
liquid
refractive index
n20/D 1.371
density
1.307 g/mL at 25 °C
functional group
fluoro
ketone
SMILES string
FCC(=O)CF
InChI
1S/C3H4F2O/c4-1-3(6)2-5/h1-2H2
InChI key
HKIPCXRNASWFRU-UHFFFAOYSA-N
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 3
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
124.0 °F - closed cup
Flash Point(C)
51.1 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service