753726
Piperidine-4-boronic acid pinacol ester hydrochloride
95%
Synonym(s):
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)piperidine hydrochloride
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C11H22BNO2 · HCl
CAS Number:
Molecular Weight:
247.57
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
95%
form
solid
mp
188-192 °C
storage temp.
2-8°C
SMILES string
Cl.CC1(C)OB(OC1(C)C)C2CCNCC2
InChI
1S/C11H22BNO2.ClH/c1-10(2)11(3,4)15-12(14-10)9-5-7-13-8-6-9;/h9,13H,5-8H2,1-4H3;1H
InChI key
DJEBJGUAIJWKGA-UHFFFAOYSA-N
Related Categories
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service