Skip to Content
Merck
All Photos(3)

Key Documents

752622

Sigma-Aldrich

Iron disilicide

greener alternative

powder, -20 mesh, 99.9% trace metals basis

Synonym(s):

Iron silicide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
FeSi2
CAS Number:
Molecular Weight:
112.02
UNSPSC Code:
12352103
NACRES:
NA.23

Quality Level

Assay

99.9% trace metals basis

form

powder

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

particle size

-20 mesh

mp

1220 °C (lit.)

greener alternative category

InChI

1S/Fe.2H2Si/h;2*1H2

InChI key

UVUDQHYBXBVHAI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Iron disilicide (FeSi2) is a direct bandgap (0.85-0.87 eV) semiconductor material that has a high optical absorption coefficient, high Seebeck coefficient, high working temperature and high resistance to oxidation.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

FeSi2 can be used in a variety of optoelectronic applications such as lithium-ion batteries, light-emitting diodes (LEDs) and solar cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nishida, I.
Physical Review. B, Condensed Matter and Materials Physics, 7, 2710-2710 (1973)
Thermoelectric properties of hot-pressed Al-and Co-doped iron disilicide materials
He Z, et al.
Journal of alloys and compounds, 438(1-2), 303-309 (2007)
Green synthesis and stable Li-storage performance of FeSi2/Si@ C nanocomposite for lithium-ion batteries
Chen Y, et al.
ACS Applied Materials & Interfaces, 4(7), 3753-3758 (2012)
Amorphous-iron disilicide: A promising semiconductor
Milosavljevic M, et al.
Applied Physics Letters, 79(10), 1438-1440 (2001)
Gross, E.; Riffel, M.; Stoehrer, U.
Journal of Materials Research, 10, 34-34 (1995)

Articles

Higher transition metal silicides are ideal for anisotropic thermoelectric conversion due to their Seebeck coefficient anisotropy and mechanical properties.

Higher transition metal silicides are ideal for anisotropic thermoelectric conversion due to their Seebeck coefficient anisotropy and mechanical properties.

Higher transition metal silicides are ideal for anisotropic thermoelectric conversion due to their Seebeck coefficient anisotropy and mechanical properties.

Higher transition metal silicides are ideal for anisotropic thermoelectric conversion due to their Seebeck coefficient anisotropy and mechanical properties.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service