750859
Ammonium-15N nitrate-18O3
98 atom % 15N, 95 atom % 18O, 98% (CP)
About This Item
Recommended Products
isotopic purity
98 atom % 15N
95 atom % 18O
Assay
98% (CP)
form
solid
bp
210 °C (lit.)
mp
169 °C (lit.)
mass shift
M+7
SMILES string
[18O-][N+]([18O-])=[18O].[15NH4+]
InChI
1S/NO3.H3N/c2-1(3)4;/h;1H3/q-1;/p+1/i2+2,3+2,4+2;1+1
InChI key
DVARTQFDIMZBAA-GNLIOFPVSA-O
Related Categories
Packaging
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Ox. Sol. 3
Storage Class Code
5.1C - Ammonium nitrate and ammonium nitrate containing preparations
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service