Skip to Content
Merck
All Photos(1)

Key Documents

750026

Sigma-Aldrich

3α,5β-Tetrahydroaldosterone

≥98% (CP)

Synonym(s):

11β,18-Epoxy-3α,18,21-trihydroxy-5β-pregnan-20-one, 5β-Pregnan-11β,18-epoxy-3α,18,21-triol-20-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H32O5
CAS Number:
Molecular Weight:
364.48
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.24

Quality Level

Assay

≥98% (CP)

form

powder

technique(s)

mass spectrometry (MS): suitable

storage temp.

−20°C

SMILES string

C[C@]12CC[C@@H](O)C[C@H]1CC[C@H]3[C@@H]4CC[C@H](C(=O)CO)[C@@]45C[C@H](O[C@@H]5O)[C@H]23

InChI

1S/C21H32O5/c1-20-7-6-12(23)8-11(20)2-3-13-14-4-5-15(16(24)10-22)21(14)9-17(18(13)20)26-19(21)25/h11-15,17-19,22-23,25H,2-10H2,1H3/t11-,12-,13+,14+,15-,17+,18-,19+,20+,21-/m1/s1

InChI key

JDEYOKPHRJVBQO-BMDKAXIVSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Egfjord et al.
Metabolism: clinical and experimental, 42(4), 470-476 (1993-04-01)
The effect of uremia on hepatic metabolism of aldosterone was studied in the isolated perfused liver of female Wistar rats. Uremia was induced by five-sixths partial nephrectomy 4 weeks before experiments. Isolated livers of normal and uremic rats were perfused
T Miyazaki et al.
Endocrinologia japonica, 32(4), 505-509 (1985-08-01)
After a large amount of aldosterone was injected into a male rabbit, urine was collected for 48 h. Separation of urinary aldosterone metabolites into monoglucosiduronate fraction and monosulphate fraction was carried out by a combination of countercurrent distribution and DEAE-Sephadex
T Miyazaki et al.
Endocrinologia japonica, 34(2), 161-169 (1987-04-01)
Analysis of urinary metabolites of [1, 2-3H]-aldosterone was performed in the male guinea-pig. Separation of urinary metabolites was carried out by countercurrent distribution followed by DEAE-Sephadex A-25 column chromatography. A major component was obtained which was both hydrolyzable with sulphatase
B J Koopman et al.
Journal of chromatography, 378(2), 283-292 (1986-06-13)
A newly devised procedure for a simultaneous determination of urinary tetrahydroaldosterone and aldosterone is described. The procedure is based on deconjugation and acetalization, followed by extraction and derivatization of the urinary compounds to their trimethylsilyl ethers and subsequent gas chromatographic-mass
S Ulick et al.
The Journal of clinical endocrinology and metabolism, 76(4), 873-878 (1993-04-01)
18-Hydroxycortisol and 18-oxocortisol have been isolated from the urine of patients with aldosterone producing adrenocortical adenomas, but not from those with idiopathic hyperaldosteronism associated with bilateral adrenal hyperplasia. These C-18 oxygenated cortisols are biosynthesized by the substitution of cortisol for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service