Skip to Content
Merck
All Photos(1)

Key Documents

742481

Sigma-Aldrich

Dimethyl 2,2′-bipyridine-4,4′-dicarboxylate

≥98.0% (HPLC)

Synonym(s):

4,4′-Bis(methoxycarbonyl)-2,2′-bipyridine, 4,4′-Dimethoxycarbonyl-2,2′-bipyridine, NSC 297840, [2,2′-Bipyridine]-4,4′-dicarboxylic acid 4,4′-dimethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H12N2O4
CAS Number:
Molecular Weight:
272.26
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (HPLC)

form

powder

composition

carbon, 61.76%
hydrogen, 4.44%
nitrogen, 10.29%

impurities

≤1.0% water

mp

208-212 °C

functional group

ester

SMILES string

COC(=O)c1ccnc(c1)-c2cc(ccn2)C(=O)OC

InChI

1S/C14H12N2O4/c1-19-13(17)9-3-5-15-11(7-9)12-8-10(4-6-16-12)14(18)20-2/h3-8H,1-2H3

InChI key

HBWBVIDKBKOVEX-UHFFFAOYSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service