Skip to Content
Merck
All Photos(1)

Key Documents

739618

Sigma-Aldrich

25-Hydroxyvitamin D3 (6,19,19-d3) solution

50 μg/mL in ethanol, 97 atom % D, 98% (CP)

Synonym(s):

25-Hydroxycholecalciferol (6,19,19-d3) solution, Deuterated 25-hydroxyvitamin D3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27D3H41O2
CAS Number:
Molecular Weight:
403.66
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.24

isotopic purity

97 atom % D

Quality Level

Assay

98% (CP)

form

liquid

concentration

50 μg/mL in ethanol

technique(s)

mass spectrometry (MS): suitable

color

colorless

application(s)

detection

mass shift

M+3

storage temp.

−20°C

SMILES string

[2H]/C([2H])=C1CC[C@H](O)CC\1=C([2H])\C=C2/CCC[C@]3(C)[C@H](CC[C@@H]23)[C@H](C)CCCC(C)(C)O

InChI

1S/C27H44O2/c1-19-10-13-23(28)18-22(19)12-11-21-9-7-17-27(5)24(14-15-25(21)27)20(2)8-6-16-26(3,4)29/h11-12,20,23-25,28-29H,1,6-10,13-18H2,2-5H3/b21-11+,22-12-/t20-,23+,24-,25+,27-/m1/s1/i1D2,12D

InChI key

JWUBBDSIWDLEOM-CMMPNOGOSA-N

Looking for similar products? Visit Product Comparison Guide

General description

25-hydroxyvitamin D3 (calcifediol) is a prohormone produced via hydroxylation of vitamin D3 (cholecalciferol) in the liver. It is a precursor for the synthesis of calcitriol or 1,25-dihydroxyvitamin D3 or [1,25(OH)2D3]. It is used as a biomarker to determine the status of vitamin D in the body.
25-Hydroxyvitamin D3 (6,19,19-d3) is a deuterated 25-hydroxyvitamin D3 wherein C-6 and C-19 protons are replaced by deuterium.

Application

25-Hydroxyvitamin D3 (6,19,19-d3) can be used as an analytical standard.

Packaging

1 mL in ampule

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

57.2 °F - closed cup

Flash Point(C)

14.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

25-Hydroxyvitamin D3, the prohormone of 1, 25-dihydroxyvitamin D3, inhibits the proliferation of primary prostatic epithelial cells
Barreto, AM et al.
Cancer Epidemiology, Biomarkers & Prevention, 9(3), 265-270 (2000)
Quantification of circulating 25-hydroxyvitamin D by liquid chromatography-tandem mass spectrometry
Vogeser M
The Journal of Steroid Biochemistry and Molecular Biology, 121(3-5), 565-573 (2010)
Jiri Adamec et al.
Journal of separation science, 34(1), 11-20 (2010-12-21)
Simultaneous and accurate measurement of vitamin D and 25-hydroxyvitamin D in biological samples is a barrier limiting our ability to define "optimal" vitamin D status. Thus, our goal was to optimize conditions and evaluate an LC-MS method for simultaneous detection

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service