Skip to Content
Merck
All Photos(1)

Key Documents

739545

Sigma-Aldrich

Pregnenolone-20,21-13C2-16,16-d2

≥98 atom %, ≥98% (CP)

Synonym(s):

3β-Hydroxy-5-pregnen-20-one-20,21-13C2-16,16-d2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C2C19D2H30O2
Molecular Weight:
320.48
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.24

isotopic purity

 13C
 D
≥98 atom %

Quality Level

Assay

≥98% (CP)

form

powder

technique(s)

mass spectrometry (MS): suitable

mass shift

M+4

storage temp.

−20°C

SMILES string

[2H]C1([2H])C[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]2(C)[C@H]1[13C]([13CH3])=O

InChI

1S/C21H32O2/c1-13(22)17-6-7-18-16-5-4-14-12-15(23)8-10-20(14,2)19(16)9-11-21(17,18)3/h4,15-19,23H,5-12H2,1-3H3/t15-,16-,17+,18-,19-,20-,21+/m0/s1/i1+1,6D2,13+1

InChI key

ORNBQBCIOKFOEO-KXHQNKTISA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antonio G Gomes de Mello Martins et al.
The Journal of steroid biochemistry and molecular biology, 174, 183-191 (2017-09-12)
CYP17A1-independent intratumoral steroid hormone synthesis is regarded as one possible explanation for resistance to treatment with the CYP17-inhibitor Abiraterone (Abi). The aim of our study was therefore to investigate the steroid metabolism of prostate cancer cells under serum starvation and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service