Skip to Content
Merck
All Photos(1)

Key Documents

739073

Sigma-Aldrich

3-(Boc-amino)azetidine

Synonym(s):

3-(Boc-amino)azetidine, tert-Butyl 3-azetidinylcarbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H16N2O2
CAS Number:
Molecular Weight:
172.22
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

solid

mp

63-72 °C

functional group

amine

SMILES string

CC(C)(C)OC(=O)NC1CNC1

InChI

1S/C8H16N2O2/c1-8(2,3)12-7(11)10-6-4-9-5-6/h6,9H,4-5H2,1-3H3,(H,10,11)

InChI key

NEMXVXVJGXZDRR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service