738778
L-Lactic acid-1-13C
≥99 atom % 13C, ≥98% (CP), ≥98% (Chiral Purity, HPLC)
Synonym(s):
(S)-2-Hydroxypropionic acid-1-13C, Sarcolactic acid-1-13C
About This Item
Recommended Products
isotopic purity
≥99 atom % 13C
Assay
≥98% (CP)
≥98% (Chiral Purity, HPLC)
form
solid
greener alternative product score
old score: 45
new score: 14
Find out more about DOZN™ Scoring
greener alternative product characteristics
Less Hazardous Chemical Syntheses
Design for Energy Efficiency
Inherently Safer Chemistry for Accident Prevention
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
technique(s)
bio NMR: suitable
mass spectrometry (MS): suitable
greener alternative category
mass shift
M+1
shipped in
dry ice
storage temp.
−20°C
SMILES string
C[C@H](O)[13C](O)=O
InChI
1S/C3H6O3/c1-2(4)3(5)6/h2,4H,1H3,(H,5,6)/t2-/m0/s1/i3+1
InChI key
JVTAAEKCZFNVCJ-NSQKCYGPSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Packaging
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Skin Corr. 1C
Supplementary Hazards
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 1
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service