Skip to Content
Merck
All Photos(1)

Key Documents

738743

Sigma-Aldrich

Polyacrylamide

average Mn 40,000

Synonym(s):

PAM

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C3H5NO)n
CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

form

powder

Quality Level

mol wt

average Mn 40,000

mp

246-250 °C

transition temp

Tm 246-250 °C

InChI

1S/C3H5NO/c1-2-3(4)5/h2H,1H2,(H2,4,5)

InChI key

HRPVXLWXLXDGHG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Proteomic analysis and lethality of the venom of Aegaeobuthus nigrocinctus, a scorpion of medical significance in the Middle East.: This study utilized polyacrylamide gel electrophoresis (PAGE) to analyze the protein composition of the venom, providing insights into its biochemical properties and potential medical applications. The results highlight the importance of PAGE in venom proteomics and its relevance in biomedical research (Borges et al., 2024).
  • One-Dimensional Acrylamide Gel Electrophoresis for Analysis of Plant Samples.: This paper describes the methodology of using PAGE for the analysis of plant proteins, demonstrating its utility in biochemistry and molecular biology studies. The technique is essential for separating and identifying proteins in complex mixtures, underscoring its significance in research and development in the life sciences (Marzban et al., 2024).
  • Protocol for isolating small cytosolic dsDNA from cultured murine cells.: The study outlines a detailed protocol incorporating PAGE for the isolation and analysis of small cytosolic dsDNA from murine cells. This method is pivotal for research in molecular and cellular biology, facilitating advancements in genetic and biomedical studies (Dai et al., 2024).
  • Comparative study of the commonly used protein quantitation assays on different Hymenoptera venoms: A fundamental aspect of Hymenoptera venom proteome analysis.: This research compares various protein quantitation assays, including PAGE, to analyze Hymenoptera venoms. The findings underscore the critical role of PAGE in proteomic analysis and its application in toxinology and biomedical research (Wanandy et al., 2024).
  • Proteomics and Its Current Application in Biomedical Area: Concise Review.: This review highlights the applications of proteomics, including PAGE, in biomedical research. It discusses the advancements and future directions in the field, emphasizing the importance of PAGE in proteomic studies and its contributions to biomedical science (Gobena et al., 2024).

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Sachio Yamamoto
Yakugaku zasshi : Journal of the Pharmaceutical Society of Japan, 132(9), 1031-1035 (2012-10-02)
Microchip electrophoresis is widely used for microfluidics and has been studied extensively over the past decade. Translation of capillary electrophoresis methods from traditional capillary systems to a microchip platform provides rapid separation and easy quantitation of sample components. However, most
D J King et al.
Reviews on environmental health, 8(1-4), 3-16 (1989-01-01)
The purpose of this paper is to present information gathered regarding, in general, the physical characteristics, and, in particular, the possible toxic nature of polyacrylamide A short discussion of the properties and toxicity of the acrylamide monomer is also included.
N V Bovin
Bioorganicheskaia khimiia, 22(9), 643-663 (1996-09-01)
The synthesis, physicochemical characteristics, and application for studying the carbohydrate-binding molecules of some analogues of human cell glycoconjugates, neoglycoconjugates, were reviewed. An approach to the synthesis of the polyacrylamide derivatives of carbohydrates based on the interaction of fully activated polyacrylic
Imran Khimji et al.
Methods (San Diego, Calif.), 64(3), 292-298 (2013-08-28)
Biosensors are devices that can provide quantitative or semi-quantitative analytical information about target molecules, where molecular recognition is based on biomolecular interactions. In recent years, DNA has emerged as a useful molecule for biosensor development since DNA can not only
E A Smith et al.
Reviews on environmental health, 9(4), 215-228 (1991-10-01)
Acrylamide is a highly water soluble vinyl monomer formed from the hydration of acrylonitrile. The major commercial use of acrylamide is the formation of polymers. In the environment acrylamide has a high mobility in soil, may travel great distances in

Articles

Highlighting existing and novel fabrication methods for both, solid and hydrogel-based scaffold for tissue engineering applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service