Skip to Content
Merck
All Photos(3)

Key Documents

738700

Sigma-Aldrich

4-Piperidinopyridine

97%

Synonym(s):

1-(4-Pyridyl)piperidine, 4-Piperidinopyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14N2
CAS Number:
Molecular Weight:
162.23
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

mp

78-83 °C

SMILES string

C1CCN(CC1)c2ccncc2

InChI

1S/C10H14N2/c1-2-8-12(9-3-1)10-4-6-11-7-5-10/h4-7H,1-3,8-9H2

InChI key

MTPBUCCXRGSDCR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Skin Corr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service