The enantiomeric purity is not tested for this product.
Please see the Sampler Certificate of Analysis for more information: https://www.sigmaaldrich.com/certificates/COFA/73/735868/735868-BULK_______MKBZ7175V_.pdf
735868
(R)-3-(Boc-amino)piperidine
97%
Synonym(s):
tert-Butyl (R)-piperidin-3-ylcarbamate, (R)-3-tert-Butoxycarbonylaminopiperidine
About This Item
Recommended Products
Quality Level
Assay
97%
form
solid
optical activity
[α]22/D +3.2°, c = 0.5 in DMF
mp
116-121 °C
functional group
amine
storage temp.
2-8°C
SMILES string
CC(C)(C)OC(=O)N[C@@H]1CCCNC1
InChI
1S/C10H20N2O2/c1-10(2,3)14-9(13)12-8-5-4-6-11-7-8/h8,11H,4-7H2,1-3H3,(H,12,13)/t8-/m1/s1
InChI key
WUOQXNWMYLFAHT-MRVPVSSYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Acute 1 - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
Please share the enantiomeric excess analysis method for this molecule
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service