732028
N-Methyl-13C, d3-diethanolamine
99 atom % 13C, 98 atom % D, 97% (CP)
Synonym(s):
2,2′-Methyl-13C,d3-iminodiethanol, N,N-Bis(2-hydroxyethyl)methyl-13C,d3-amine, DMEA-methyl-13C,d3
About This Item
Recommended Products
isotopic purity
99 atom % 13C
98 atom % D
Assay
97% (CP)
form
liquid
bp
246-248 °C (lit.)
density
1.072 g/mL at 25 °C
mass shift
M+4
SMILES string
[2H][13C]([2H])([2H])N(CCO)CCO
InChI
1S/C5H13NO2/c1-6(2-4-7)3-5-8/h7-8H,2-5H2,1H3/i1+1D3
InChI key
CRVGTESFCCXCTH-KQORAOOSSA-N
Related Categories
Packaging
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
260.6 °F - closed cup
Flash Point(C)
127 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service